2,1,3-Benzothiadiazole-4-sulphonyl chloride
Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Benzo[c][1,2,5]thiadiazole-4-sulphonyl chloride
FLUO005896-2.5GEA
0
GBP
FLUO005896-2.5G
2,1,3-Benzothiadiazole-4-sulphonyl chloride
2,1,3-Benzothiadiazole-4-sulfonyl chloride
Formula:
C₆H₃ClN₂O₂S₂ MW: 234.68 g/mol |
MDL Number:
MFCD00068049 CAS Number: 73713-79-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...