Thiophene-3-carboxaldoxime ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L13114.03EA
26.4
GBP
L13114.03
L13114.06
Thiophene-3-carboxaldoxime ≥98%
Thiophene-3-carboxaldoxime
Formula:
C₅H₅NOS MW: 127.17 g/mol Melting Pt: 121…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151853 CAS Number: 42466-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...