4-Nitro-2,3-lutidine-N-oxide ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2,3-Dimethyl-4-nitropyridine 1-oxide
, 2,3-Dimethyl-4-nitropyridine N-oxide
L19457.03EA
16.6
GBP
L19457.03
4-Nitro-2,3-lutidine-N-oxide ≥97%
4-Nitro-2,3-lutidine N-oxide
Formula:
C₇H₈N₂O₂ MW: 168.15 g/mol Melting Pt: 92…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065172 CAS Number: 37699-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...