6-Methylnicotinamide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
6-Methylpyridine-3-carboxamide
M-Clarity™ quality level = MQ100
SIAL284769-5GEA
189
GBP
SIAL284769-5G
6-Methylnicotinamide, Sigma-Aldrich®
6-Methylnicotinamide
Formula:
C₇H₈N₂O MW: 136.15 g/mol |
MDL Number:
MFCD00006342 CAS Number: 6960-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...