2-Chloro-3-methyl-4-nitropyridine-N-oxide
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBPY-7979-1GEA
126.62
GBP
COBBPY-7979-1G
COBBPY-7979-5G
2-Chloro-3-methyl-4-nitropyridine-N-oxide
2-Chloro-3-methyl-4-nitropyridine N-oxide
Formula:
C₆H₅ClN₂O₃ |
MDL Number:
MFCD01194031 CAS Number: 60323-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...