2-Chloro-6-(trifluoromethyl)nicotinic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-6-(trifluoromethyl)pyridine-3-carboxylic acid

SIAL708208-250MGEA 72 GBP
SIAL708208-250MG
2-Chloro-6-(trifluoromethyl)nicotinic acid, Sigma-Aldrich®
2-Chloro-6-(trifluoromethyl)nicotinic acid
Formula: C₇H₃ClF₃NO₂
MW: 225.56 g/mol
MDL Number: MFCD01862658
CAS Number: 280566-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR