4-Cyanopyridine-N-oxide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Cyanopyridine 1-oxide
M-Clarity™ quality level = MQ100
SIAL142352-5GEA
68.5
GBP
SIAL142352-5G
4-Cyanopyridine-N-oxide, Sigma-Aldrich®
4-Cyanopyridine N-oxide
Formula:
C₆H₄N₂O MW: 120.1088 g/mol |
MDL Number:
MFCD00006205 CAS Number: 14906-59-3 EINECS: 238-974-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...