3-Chloro-5-fluoropyridine-N-oxide

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

COBBPY-1360-1GEA 114.75 GBP
COBBPY-1360-1G
3-Chloro-5-fluoropyridine-N-oxide
3-Chloro-5-fluoropyridine N-oxide
Formula: C₅H₃ClFNO
MDL Number: MFCD16036157
CAS Number: 1221793-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR