3-Bromo-5-fluoropyridine-N-oxide

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

COBBPY-1358-5GEA 300.73 GBP
COBBPY-1358-5G
3-Bromo-5-fluoropyridine-N-oxide
3-Bromo-5-fluoropyridine N-oxide
Formula: C₅H₃BrFNO
MDL Number: MFCD16036155
CAS Number: 1221793-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR