3-Bromo-5-chloropyridine-N-oxide
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBPY-1357-1GEA
0
GBP
COBBPY-1357-1G
3-Bromo-5-chloropyridine-N-oxide
3-Bromo-5-chloropyridine N-oxide
Formula:
C₅H₃BrClNO |
MDL Number:
MFCD16036154 CAS Number: 1221793-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...