1,8-Bis(bromomethyl)naphthalene, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
SIAL246921-250MGEA
145
GBP
SIAL246921-250MG
1,8-Bis(bromomethyl)naphthalene, Sigma-Aldrich®
1,8-Bis(bromomethyl)naphthalene
Formula:
C₁₂H₁₀Br₂ |
MDL Number:
MFCD00010251 CAS Number: 2025-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...