4-Bromo-1-naphthaleneboronic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(4-Bromonaphthalen-1-yl)boronic acid
COBBBB-9079-1GEA
0
GBP
COBBBB-9079-1G
4-Bromo-1-naphthaleneboronic acid
4-Bromo-1-naphthaleneboronic acid
Formula:
C₁₀H₈BBrO₂ MW: 250.88 g/mol |
MDL Number:
MFCD08701764 CAS Number: 145965-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...