Diiron phosphide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
SIAL691658-5GEA
110
GBP
SIAL691658-5G
Diiron phosphide, Sigma-Aldrich®
Diiron phosphide
Formula:
Fe₂P |
CAS Number:
1310-43-6 EINECS: 215-178-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...