Dimanganese antimonide ≥99.5% (metals basis)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Manganese antimonide
36390.06EA
67.7
GBP
36390.06
36390.14
Dimanganese antimonide ≥99.5% (metals basis)
Dimanganese antimonide
Formula:
Mn₂Sb MW: 231.63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00151354 CAS Number: 12032-97-2 EINECS: 234-784-2 UN: 1549 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...