Manganese trialuminide ≥99.5% (metals basis)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Manganese aluminide
36384.14EA
271
GBP
36384.14
36384.06
Manganese trialuminide ≥99.5% (metals basis)
Manganese trialuminide
Formula:
Al₃Mn MW: 135.88 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00213943 CAS Number: 12253-13-3 EINECS: 235-492-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...