Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triytterbium pentasilicide ≥99.9% (metals basis excluding Ta) (max. 0,4% Ta)

Supplier: Alfa Aesar

41523.06 41523.03
Triytterbium pentasilicide ≥99.9% (metals basis excluding Ta) (max. 0,4% Ta)


Formula: Si₅Yb₃
MDL Number: MFCD00148826
CAS Number: 12039-89-3
EINECS: 234-910-6
UN: 0000
Order Now

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty