Magnesium wolframate ≥99% (metals basis)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Magnesium tungsten oxide
, Magnesium tungstate
14023.14EA
50.6
GBP
14023.14
14023.22
Magnesium wolframate ≥99% (metals basis)
Magnesium wolframate
Formula:
MgO₄W MW: 272.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00016209 CAS Number: 13573-11-0 EINECS: 236-999-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...