1-Allylcyclohexanol ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
A19066.88EA
36.2
GBP
A19066.88
A19066.09
1-Allylcyclohexanol ≥97%
1-Allylcyclohexanol
Formula:
C₉H₁₆O MW: 140.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00021403 CAS Number: 1123-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...