Cyclopropylboronic acid pinacol ester
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
COBBPN-2007-25GEA
75.18
GBP
COBBPN-2007-25G
COBBPN-2007-1G
COBBPN-2007-5G
Cyclopropylboronic acid pinacol ester
Cyclopropylboronic acid pinacol ester
Formula:
C₉H₁₇BO₂ MW: 168.04 g/mol |
MDL Number:
MFCD05663847 CAS Number: 126689-01-8 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...