3-(Benzyloxy)-6-bromo-2-fluoroanisole

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

COBBOT-1228-1GEA 0 GBP
COBBOT-1228-1G
3-(Benzyloxy)-6-bromo-2-fluoroanisole
3-(Benzyloxy)-6-bromo-2-fluoroanisole
Formula: C₁₄H₁₂BrFO₂
MDL Number: MFCD16618958
CAS Number: 1228957-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR