2-(Methoxycarbonyl)-4-(trifluoromethyl)phenylboronic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBBB-5304-1GEA
0
GBP
COBBBB-5304-1G
COBBBB-5304-100M
COBBBB-5304-500M
2-(Methoxycarbonyl)-4-(trifluoromethyl)phenylboronic acid
2-(Methoxycarbonyl)-4-(trifluoromethyl)phenylboronic acid
Formula:
C₉H₈BF₃O₄ |
MDL Number:
MFCD11504848 CAS Number: 1072951-42-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...