2-(Methoxycarbonyl)-3-(trifluoromethyl)phenylboronic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBBB-5282-1GEA
276.99
GBP
COBBBB-5282-1G
COBBBB-5282-250M
2-(Methoxycarbonyl)-3-(trifluoromethyl)phenylboronic acid
2-(Methoxycarbonyl)-3-(trifluoromethyl)phenylboronic acid
Formula:
C₉H₈BF₃O₄ |
MDL Number:
MFCD12025963 CAS Number: 1150114-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...