2,5-Bis(chloromethyl)-4-(3,7-dimethyloctyloxy)anisole, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIAL546445-1GEA
585
GBP
SIAL546445-1G
2,5-Bis(chloromethyl)-4-(3,7-dimethyloctyloxy)anisole, Sigma-Aldrich®
2,5-Bis(chloromethyl)-4-(3,7-dimethyloctyloxy)anisole
Formula:
C₁₉H₃₀Cl₂O₂ |
MDL Number:
MFCD03427076 CAS Number: 174097-32-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...