2,5-Bis(bromomethyl)-4-(3,7-dimethyloctyloxy)anisole, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIAL546453-1GEA
506
GBP
SIAL546453-1G
2,5-Bis(bromomethyl)-4-(3,7-dimethyloctyloxy)anisole, Sigma-Aldrich®
2,5-Bis(bromomethyl)-4-(3,7-dimethyloctyloxy)anisole
Formula:
C₁₉H₃₀Br₂O₂ |
MDL Number:
MFCD03094030 CAS Number: 287919-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...