4-Toluenethiol sodium salt, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Methylbenzenethiol sodium salt
SIAL513911-10GEA
226
GBP
SIAL513911-10G
4-Toluenethiol sodium salt, Sigma-Aldrich®
4-Toluenethiol sodium salt
Formula:
C₇H₇NaS |
MDL Number:
MFCD00066208 CAS Number: 10486-08-5 EINECS: 234-000-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...