2-(p-Toluenesulphonyl)ethanol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Tosylethanol
M-Clarity™ quality level = MQ100
SIAL446386-10GEA
300
GBP
SIAL446386-10G
2-(p-Toluenesulphonyl)ethanol, Sigma-Aldrich®
2-(p-Toluenesulphonyl)ethanol
Formula:
C₉H₁₂O₃S MW: 200.2600 g/mol |
MDL Number:
MFCD00041757 CAS Number: 22381-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...