Tolbutamide
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PROOICRS0431EA
0
GBP
PROOICRS0431
Tolbutamide
Tolbutamide
Formula:
C₁₂H₁₈N₂O₃S MW: 270,35 |
MDL Number:
MFCD00027169 CAS Number: 64-77-7 EINECS: 200-594-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...