3,5-Dichloro-2-methylphenylboronic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBBB-5026-1GEA
0
GBP
COBBBB-5026-1G
3,5-Dichloro-2-methylphenylboronic acid
3,5-Dichloro-2-methylphenylboronic acid
Formula:
C₇H₇BCl₂O₂ |
MDL Number:
MFCD09800882 CAS Number: 957120-97-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...