2-(Bromomethyl)-4-fluorophenylboronic acid pinacol ester
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-(2-(Bromomethyl)-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
COBBPN-2706-1GEA
91.01
GBP
COBBPN-2706-1G
COBBPN-2706-5G
2-(Bromomethyl)-4-fluorophenylboronic acid pinacol ester
2-(Bromomethyl)-4-fluorophenylboronic acid pinacol ester
Formula:
C₁₃H₁₇BBrFO₂ MW: 314.9863 g/mol |
MDL Number:
MFCD06659927 CAS Number: 850567-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...