3-(Hydroxymethyl)phenylboronic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-(Hydroxymethyl)benzeneboronic acid
M-Clarity™ quality level = MQ100
SIAL512834-10GEA
84.2
GBP
SIAL512834-10G
3-(Hydroxymethyl)phenylboronic acid, Sigma-Aldrich®
3-(Hydroxymethyl)phenylboronic acid
Formula:
C₇H₉BO₃ MW: 151.96 g/mol |
MDL Number:
MFCD01317846 CAS Number: 87199-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...