2-Formyl-5-methylphenylboronic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBBB-8334-1GEA
43.53
GBP
COBBBB-8334-250M
COBBBB-8334-1G
2-Formyl-5-methylphenylboronic acid
2-Formyl-5-methylphenylboronic acid
Formula:
C₈H₉BO₃ MW: 163.9663 g/mol |
MDL Number:
MFCD01319009 CAS Number: 40138-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...