3-[(Trimethylsilyl)ethynyl]benzaldehyde, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIAL709727-250MGEA
101
GBP
SIAL709727-250MG
3-[(Trimethylsilyl)ethynyl]benzaldehyde, Sigma-Aldrich®
3-[(Trimethylsilyl)ethynyl]benzaldehyde
Formula:
C₁₂H₁₄OSi |
MDL Number:
MFCD06245348 CAS Number: 77123-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...