2-Mercapto-4-phenylthiazole, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Phenylthiazole-2-thiol
M-Clarity™ quality level = MQ100
SIAL632570-5GEA
122
GBP
SIAL632570-5G
2-Mercapto-4-phenylthiazole, Sigma-Aldrich®
2-Mercapto-4-phenylthiazole
Formula:
C₉H₇NS₂ MW: 193.30 g/mol |
MDL Number:
MFCD00014508 CAS Number: 2103-88-0 EINECS: 218-274-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...