2-Amino-4-(2,4-difluorophenyl)thiazole, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
SIAL683280-1GEA
94.3
GBP
SIAL683280-1G
2-Amino-4-(2,4-difluorophenyl)thiazole, Sigma-Aldrich®
2-Amino-4-(2,4-difluorophenyl)thiazole
Formula:
C₉H₆F₂N₂S MW: 212.22 g/mol |
MDL Number:
MFCD00472891 CAS Number: 105512-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...