2-Amino-4-(4-bromophenyl)thiazole, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
SIAL534129-5GEA
73.8
GBP
SIAL534129-5G
2-Amino-4-(4-bromophenyl)thiazole, Sigma-Aldrich®
2-Amino-4-(4-bromophenyl)thiazole
Formula:
C₉H₇BrN₂S MW: 255.1343 g/mol |
MDL Number:
MFCD00051953 CAS Number: 2103-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...