2,4-Diaminoanisole sulphate hydrate

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

MOLE46715302-500GEA 0 GBP
MOLE46715302-500G
2,4-Diaminoanisole sulphate hydrate
2,4-Diaminoanisole sulphate hydrate
Formula: C₇H₁₂N₂O₅S·H₂O
MDL Number: MFCD00012978
CAS Number: 123333-56-2
EINECS: 254-323-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR