3-Amino-4-methoxybenzenesulphonic acid
Supplier: ABCR GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABCRAB136696.0500EA
0
GBP
ABCRAB136696.0500
3-Amino-4-methoxybenzenesulphonic acid
3-Amino-4-methoxybenzenesulphonic acid
Formula:
C₇H₉NO₄S MW: 203.2157 g/mol |
MDL Number:
MFCD00035759 CAS Number: 98-42-0 EINECS: 202-667-5 UN: 2585 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...