4-Phenoxybenzonitrile, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
SIAL527769-1GEA
60.6
GBP
SIAL527769-1G
4-Phenoxybenzonitrile, Sigma-Aldrich®
4-Phenoxybenzonitrile
Formula:
C₁₃H₉NO MW: 195.22 g/mol |
MDL Number:
MFCD00017346 CAS Number: 3096-81-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...