4-Phenoxybenzonitrile
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBCA-4398-1GEA
0
GBP
COBBCA-4398-1G
COBBCA-4398-100G
COBBCA-4398-25G
COBBCA-4398-5G
4-Phenoxybenzonitrile
4-Phenoxybenzonitrile
Formula:
C₁₃H₉NO MW: 195.22 g/mol |
MDL Number:
MFCD00017346 CAS Number: 3096-81-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...