4,5-Dimethoxy-2-nitrobenzonitrile, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
SIAL650102-5GEA
40.6
GBP
SIAL650102-5G
SIAL650102-25G
4,5-Dimethoxy-2-nitrobenzonitrile, Sigma-Aldrich®
4,5-Dimethoxy-2-nitrobenzonitrile
Formula:
C₉H₈N₂O₄ |
MDL Number:
MFCD00017564 CAS Number: 102714-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...