4-Chloro-N-cyclohexylbenzamide
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBCA-4547-25GEA
0
GBP
COBBCA-4547-25G
COBBCA-4547-1G
COBBCA-4547-5G
4-Chloro-N-cyclohexylbenzamide
4-Chloro-N-cyclohexylbenzamide
Formula:
C₁₃H₁₆ClNO MW: 237.73 g/mol |
MDL Number:
MFCD00029412 CAS Number: 57707-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...