3-(Pentafluorothio)benzaldehyde ≥97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
H35195.03EA
38.4
GBP
H35195.03
3-(Pentafluorothio)benzaldehyde ≥97%
3-(Pentafluorothio)benzaldehyde
Formula:
C₇H₅F₅OS MW: 232.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03788517 CAS Number: 401892-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...