2-Bromo-N-pentylbenzenesulphonamide
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBHC-6155-1GEA
47.48
GBP
COBBHC-6155-1G
COBBHC-6155-5G
2-Bromo-N-pentylbenzenesulphonamide
2-Bromo-N-pentylbenzenesulphonamide
Formula:
C₁₁H₁₆BrNO₂S |
MDL Number:
MFCD09800967 CAS Number: 951883-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...