2-Bromo-N-hexylbenzenesulphonamide
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBHC-6150-5GEA
142.45
GBP
COBBHC-6150-5G
COBBHC-6150-1G
2-Bromo-N-hexylbenzenesulphonamide
2-Bromo-N-hexylbenzenesulphonamide
Formula:
C₁₂H₁₈BrNO₂S |
MDL Number:
MFCD09800964 CAS Number: 951883-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...