2-Bromo-N-ethylbenzenesulphonamide
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBHC-6133-1GEA
47.48
GBP
COBBHC-6133-1G
COBBHC-6133-5G
2-Bromo-N-ethylbenzenesulphonamide
2-Bromo-N-ethylbenzenesulphonamide
Formula:
C₈H₁₀BrNO₂S |
MDL Number:
MFCD08873247 CAS Number: 169189-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...