2-Bromo-N-isopropylbenzenesulphonamide
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBHC-6128-5GEA
118.71
GBP
COBBHC-6128-5G
COBBHC-6128-1G
2-Bromo-N-isopropylbenzenesulphonamide
2-Bromo-N-isopropylbenzenesulphonamide
Formula:
C₉H₁₂BrNO₂S |
MDL Number:
MFCD09800949 CAS Number: 951883-94-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...