4-Bromo-2-(cyclopentyloxy)anisole
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBOT-1247-5GEA
379.87
GBP
COBBOT-1247-5G
COBBOT-1247-1G
4-Bromo-2-(cyclopentyloxy)anisole
4-Bromo-2-(cyclopentyloxy)anisole
Formula:
C₁₂H₁₅BrO₂ |
MDL Number:
MFCD00673135 CAS Number: 138509-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...