2-Bromo-4-(trifluoromethoxy)thioanisole

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

COBBOT-0575-1GEA 0 GBP
COBBOT-0575-1G COBBOT-0575-5G
2-Bromo-4-(trifluoromethoxy)thioanisole
2-Bromo-4-(trifluoromethoxy)thioanisole
Formula: C₈H₆BrF₃OS
MDL Number: MFCD09801038
CAS Number: 951884-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR