3-Bromo-5-(trifluoromethoxy)benzonitrile 97%, JRD

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

H31778.03EA 70.6 GBP
H31778.03 H31778.06
3-Bromo-5-(trifluoromethoxy)benzonitrile 97%, JRD
3-Bromo-5-(trifluoromethoxy)benzonitrile
Formula: C₈H₃BrF₃NO
MW: 266.01 g/mol
Melting Pt: 35…38 °C
Storage Temperature: Ambient
MDL Number: MFCD08458074
CAS Number: 914635-52-2
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR