Tribromsalan
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EHERC17666500EA
229
GBP
EHERC17666500
Tribromsalan
Tribromsalan
Formula:
C₁₃H₈Br₃NO₂ |
MDL Number:
MFCD00045740 CAS Number: 87-10-5 EINECS: 201-723-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...